Skip to Content
Merck
All Photos(2)

Key Documents

36503

Supelco

EPN

PESTANAL®, analytical standard

Synonym(s):

O-Ethyl O-(4-nitrophenyl) phenylphosphonothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14NO4PS
CAS Number:
Molecular Weight:
323.30
Beilstein:
2542580
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(Oc1ccc(cc1)[N+]([O-])=O)c2ccccc2

InChI

1S/C14H14NO4PS/c1-2-18-20(21,14-6-4-3-5-7-14)19-13-10-8-12(9-11-13)15(16)17/h3-11H,2H2,1H3

InChI key

AIGRXSNSLVJMEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EPN belongs to the class of organophosphorus insecticides, which acts as an acetylcholinesterase inhibitor[1] and is used for the control of pests in agricultural crops.[2]

Application

EPN may be used as an analytical reference standard for the determination of the analyte in biological samples[1][3] using various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

International Pesticide Directory.
McDonald DP.
International Pest Control, 13 (1995)
Martin Ochoa et al.
Environmental pollution (Barking, Essex : 1987), 260, 114050-114050 (2020-02-12)
Rice is the world's most consumed and in-demand grain. Ecuador is one of the main rice-consuming countries in Latin America, with an average per capita consumption of 53.2 kg per year. Rice cultivation takes place under flooding conditions, which favors the
Linus Holm et al.
Scientific reports, 9(1), 11265-11265 (2019-08-04)
Humans often appear to desire information for its own sake, but it is presently unclear what drives this desire. The important role that resolving uncertainty plays in stimulating information seeking has suggested a tight coupling between the intrinsic motivation to
Mareike Bayer et al.
Social cognitive and affective neuroscience, 12(9), 1470-1479 (2017-05-26)
Emotional stimuli attract attention and lead to increased activity in the visual cortex. The present study investigated the impact of personal relevance on emotion processing by presenting emotional words within sentences that referred to participants' significant others or to unknown
Yuming Chen et al.
Frontiers in psychology, 9, 1127-1127 (2018-07-20)
Directed attention is a fundamental mental resource for voluntarily managing the focus and direction of cognitive resources. The present study investigated how processing of unpleasant and neutral images is affected by emotion and previous directed attention. The results showed that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service