Skip to Content
Merck
All Photos(1)

Key Documents

36357

Supelco

Malondialdehyde tetrabutylammonium salt

analytical standard

Synonym(s):

Malonaldehyde tetrabutylammonium salt, Tetrabutylammonium malondialdehyde enolate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€63.40

€63.40


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€63.40

About This Item

Linear Formula:
OCHCH=CHO[N(CH2CH2CH2CH3)4]
CAS Number:
Molecular Weight:
313.52
Beilstein:
5191808
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€63.40


Estimated to ship on02 June 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (NT)

shelf life

limited shelf life, expiry date on the label

mp

126-132 °C

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)\C=C\[O-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.C3H4O2/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;4-2-1-3-5/h5-16H2,1-4H3;1-4H/q+1;/p-1/b;2-1+

InChI key

GFXLQCGVBUWBHF-WLHGVMLRSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gonzalo I Gómez et al.
International journal of molecular sciences, 19(7) (2018-06-27)
Boldine, a major aporphine alkaloid found in the Chilean boldo tree, is a potent antioxidant. Oxidative stress plays a detrimental role in the pathogenesis of kidney damage in renovascular hypertension (RVH). The activation of the renin-angiotensin system (RAS) is crucial
Kamolchanok Umnajkitikorn et al.
Plants (Basel, Switzerland), 10(2) (2021-03-07)
Under water deficit conditions, the essential macronutrient nitrogen becomes limited as a result of reduced dissolved nitrogen and root nitrogen uptake. An elevated nitrogen level might be able to mitigate these effects, integrated with the idea of using nitric oxide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service