Skip to Content
Merck
All Photos(1)

Key Documents

383538

Sigma-Aldrich

Malonaldehyde bis(phenylimine) monohydrochloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€91.80
100 G
€297.00

€91.80


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€91.80
100 G
€297.00

About This Item

Linear Formula:
CH2(CH=NC6H5)2·HCl
Molecular Weight:
258.75
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€91.80


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

97%

form

powder

mp

218 °C (dec.) (lit.)

solubility

methanol: 2.5%, clear, yellow to orange

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl.C(\C=N\c1ccccc1)\C=N\c2ccccc2

InChI

1S/C15H14N2.ClH/c1-3-8-14(9-4-1)16-12-7-13-17-15-10-5-2-6-11-15;/h1-6,8-13H,7H2;1H/b16-12+,17-13+;

InChI key

FLMLOZWXXPLHIE-IEEHZGKWSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service