Skip to Content
Merck
All Photos(1)

Key Documents

34138

Supelco

Neosolaniol solution

~100 μg/mL in acetonitrile, analytical standard

Synonym(s):

4β,15-Diacetoxy-3α,8α-dihydroxy-12,13-epoxytrichothec-9-ene, 8-Hydroxydiacetoxyscirpenol solution, NEO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O8
CAS Number:
Molecular Weight:
382.40
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

~100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CC(=O)C[C@@H]1[C@@H](O)[C@H]2O[C@@H]3C=C(C)[C@@H](O)C[C@]3(COC(C)=O)[C@]1(C)[C@@]24CO4

InChI

1S/C19H26O8/c1-9-5-13-18(6-12(9)22,7-24-10(2)20)17(4)15(26-11(3)21)14(23)16(27-13)19(17)8-25-19/h5,12-16,22-23H,6-8H2,1-4H3/t12-,13?,14+,15+,16+,17+,18+,19-/m1/s1

InChI key

TVZHDVCTOCZDNE-VUQWXZGYSA-N

General description

Neosolaniol is a group A trichothecene mycotoxin produced by fungi of the Fusarium genus, found in grains used for livestock as well as human consumption products such as bread and cereals.[1][2]

Application

Neosolaniol solution may be used as an analytical reference standard for the estimation of the analyte in human urine samples using gas chromatography coupled to tandem mass spectrometry (GC-MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥96.0% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justinas Sakas et al.
Faraday discussions, 218(0), 191-201 (2019-06-04)
For the structure determination of molecules in mixtures using NMR spectroscopy, the dispersion of 13C chemical shifts provides much needed separation of resonances in the indirectly detected dimension of 2D heterocorrelated NMR experiments. This separation is crucial for establishing networks
Zhiyu Xu et al.
ACS infectious diseases, 3(9), 653-665 (2017-08-02)
Aminoglycoside N-acetyltransferases (AACs) confer resistance against the clinical use of aminoglycoside antibiotics. The origin of AACs can be traced to environmental microbial species representing a vast reservoir for new and emerging resistance enzymes, which are currently undercharacterized. Here, we performed
Merijn van de Laar et al.
Psychology, health & medicine, 22(8), 902-910 (2017-01-31)
Insomnia is a too general term for various subtypes that might have different etiologies and therefore require different types of treatment. In this explorative study we used cluster analysis to distinguish different phenotypes in 218 patients with insomnia, taking into
Patrick MacDonald Fisher et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 42(2), 446-453 (2016-10-21)
Women are at relatively greater lifetime risk for depression than men. This elevated risk in women is partly due to heightened risk during time periods characterized by marked fluctuations in sex hormones, including postpartum and perimenopausal periods. How sex hormone
Pengyang Zhou et al.
Cancer letters, 385, 39-45 (2016-11-09)
Tumor-associated-fibroblasts (TAFs) are the most important host cells in the stroma and take part in extracellular matrix construction and cancer colony development. During cancer colonization, seed cells from primary tumor can reconstruct the microenvironment by recruiting circulating cancer cells and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service