Skip to Content
Merck
All Photos(1)

Documents

34062BC

Millipore

Transia® Plate Washing Buffer, 20X

BioControl, For use with Transia® Plate, volume 800 mL

Synonym(s):

Wash buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NA.85

form

liquid

packaging

pkg of 1 pieces

manufacturer/tradename

BioControl Systems TRANSIA® SA0180

storage condition

protect from light

technique(s)

ELISA: suitable

volume

800 mL

application(s)

food and beverages

storage temp.

2-8°C

General description

Transia® Plate Washing Buffer is designed to be used with the Transia® detection system. Transia® PLATE is a sandwich enzyme immunoassay to detect pathogens in food, feed, and environmental samples. The test utilizes specific proprietary antibodies to create an antigen-antibody complex. The addition of substrate initiates a colour change reaction in the presence of the antigen-antibody complex, which is read with a microplate reader. The buffer is used between the incubation steps to remove any background samples or reagents during the assay procedure

Application

Transia® Plate Washing Buffer is used between the incubation steps to remove any background sample or reagents during the assay procedure of detecting pathogens by ELISA based method in food, feed, and environmental samples.

Legal Information

TRANSIA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service