Skip to Content
Merck
All Photos(1)

Key Documents

33987

Supelco

Olaquindox

VETRANAL®, analytical standard

Synonym(s):

N-(2-Hydroxyethyl)-3-methyl-2-quinoxalinecarboxamide 1,4-dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€115.00

€115.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€115.00

About This Item

Empirical Formula (Hill Notation):
C12H13N3O4
CAS Number:
Molecular Weight:
263.25
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€115.00


Estimated to ship on22 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cc1c(C(=O)NCCO)[n+]([O-])c2ccccc2[n+]1[O-]

InChI

1S/C12H13N3O4/c1-8-11(12(17)13-6-7-16)15(19)10-5-3-2-4-9(10)14(8)18/h2-5,16H,6-7H2,1H3,(H,13,17)

InChI key

TURHTASYUMWZCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Olaquindox is an antibiotic veterinary drug, used as a growth promoter for pig farming.[1]

Application

Olaquindox may be used as a reference standard in the determination of olaquindox in feed samples using matrix solid-phase dispersion extraction (MSPDE) and hydrophilic interaction ultra-high-pressure liquid chromatography (HILIC-UHPLC) with photodiode array detector.[2][3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wentao Guo et al.
PloS one, 7(8), e43322-e43322 (2012-09-07)
Quinoxaline 1, 4-dioxides (QdNOs) has been used in animals as antimicrobial agents and growth promoters for decades. However, the resistance to QdNOs in pathogenic bacteria raises worldwide concern but it is barely known. To explore the molecular mechanism involved in
Zhao-Ying Liu et al.
Toxicology letters, 200(1-2), 24-33 (2010-10-27)
Olaquindox is a growth-promoting feed additive for food-producing animals. Its toxicities were reported to be closely related to the metabolism. To provide the interpretation of toxicities in animals, this study explored the metabolism of olaquindox in rats, chickens and pigs
Xian-Ju Huang et al.
Chemico-biological interactions, 185(3), 227-234 (2010-03-02)
Quinoxaline 1,4-dioxides (QdNOs) derivatives, the potent synthetic antibacterial group used in food-producing animals, are assumed to have pro-oxidant properties. However, how oxidative stress mediated their adrenal toxicity is far from clear. The aim of this study was to assess the
Biodegradability of metronidazole, olaquindox, and tylosin and formation of tylosin degradation products in aerobic soil--manure slurries
Ingerslev F and Halling-Sorensen B
Ecotoxicology and Environmental Safety, 48(3), 311-320 (2001)
Xu Wang et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(5), 1068-1079 (2011-01-27)
To investigate the teratogenic potential and reproductive toxicity of cyadox, a growth promoting agent, Wistar rats (F(0)) were fed with diets containing cyadox (0, 50, 150 and 2500 mg/kg) or olaquindox (150 mg/kg), approximately equivalent to cyadox 5, 15, 250

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service