Skip to Content
Merck
All Photos(2)

Key Documents

32133

Supelco

Desoxycarbadox

VETRANAL®, analytical standard

Synonym(s):

2-(2-Quinoxalinylmethylene)hydrazinecarboxylic acid methyl ester, Methyl 3-(2-quinoxalinylmethylene)carbazate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10N4O2
CAS Number:
Molecular Weight:
230.22
Beilstein:
799004
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

COC(=O)N\N=C\c1cnc2ccccc2n1

InChI

1S/C11H10N4O2/c1-17-11(16)15-13-7-8-6-12-9-4-2-3-5-10(9)14-8/h2-7H,1H3,(H,15,16)/b13-7+

InChI key

SOEMFPLGOUBPJQ-NTUHNPAUSA-N

General description

Desoxycarbadox is a metabolite of the antimicrobial growth promoting agent, carbodox.

Application

Desoxycarbadox may be used as a reference standard in the determination of desoxycarbadox in porcine tissue samples using liquid chromatography coupled with tandem mass spectrometry (LC-MS-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G J de Graaf et al.
The Veterinary quarterly, 10(1), 34-41 (1988-01-01)
Concentrations of carbadox and a first metabolite, desoxycarbadox, were measured in contents of the porcine gastrointestinal tract after in-feed administration of carbadox in therapeutic dosages (100-150 ppm). The levels of carbadox in the relevant parts of the gastrointestinal tract were
Antimicrobial Therapy in Veterinary Medicine (2013)
Residues of carbadox metabolites in edible pork products.
A J Baars et al.
Archives of toxicology. Supplement. = Archiv fur Toxikologie. Supplement, 14, 288-292 (1991-01-01)
A I MacIntosh et al.
Journal - Association of Official Analytical Chemists, 68(4), 665-671 (1985-07-01)
A liquid chromatographic method was used to monitor a depletion study of carbadox (and its most important metabolite, desoxycarbadox) in young pigs fed carbadox-treated rations for 1 week. Carbadox was found in blood (20 ppb), blood serum (26 ppb), and
A I MacIntosh et al.
Journal - Association of Official Analytical Chemists, 67(5), 958-962 (1984-09-01)
A liquid chromatographic (LC) method has been developed for the determination of carbadox, desoxycarbadox, and nitrofurazones in the 10-40 ppb range in pork muscle, liver, and kidney tissues. Tissues were homogenized in absolute ethanol, and the homogenates were treated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service