Skip to Content
Merck
All Photos(1)

Documents

33767

Supelco

(E)-Mevinphos solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Synonym(s):

(E)-3-(Dimethoxyphosphinyloxy)-2-butenoic acid methyl ester, cis-Mevinphos solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13O6P
CAS Number:
Molecular Weight:
224.15
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

COC(=O)\C=C(/C)OP(=O)(OC)OC

InChI

1S/C7H13O6P/c1-6(5-7(8)10-2)13-14(9,11-3)12-4/h5H,1-4H3/b6-5+

InChI key

GEPDYQSQVLXLEU-AATRIKPKSA-N

General description

(E)-Mevinphos is the E-isomer of the organophosphorus insecticide, mevinphos, used to control chewing and sucking insect pests in crop production. Its mode of action involves the inhibition of acetylcholinesterase activity in insect pests.

Application

(E)-Mevinphos may be used as an analytical reference standard for the determination of the analyte in groundwater samples using automated online liquid-solid extraction followed by liquid chromatography-atmospheric pressure chemical ionization mass spectrometry (LC/APCI-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Part 2: Insecticides and Fungicides.
Metabolic Pathways of Agrochemicals: Herbicides and plant growth regulators;, 68(15), 2464-2470 (2007)
Determination of parts per trillion levels of organophosphorus pesticides in groundwater by automated on-line liquid- solid extraction followed by liquid chromatography/atmospheric pressure chemical ionization mass spectrometry using positive and negative ion modes of operation.
Lacorte S and Barcelo D
Analytical Chemistry, 68(15), 2464-2470 (1996)

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service