Skip to Content
Merck
All Photos(1)

Key Documents

33587

Supelco

Fonofos solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15OPS2
CAS Number:
Molecular Weight:
246.33
Beilstein:
1958949
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

CCOP(=S)(CC)Sc1ccccc1

InChI

1S/C10H15OPS2/c1-3-11-12(13,4-2)14-10-8-6-5-7-9-10/h5-9H,3-4H2,1-2H3

InChI key

KVGLBTYUCJYMND-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toxicity of aldicarb and fonofos to the early-life-stage of the fathead minnow.
Q H Pickering et al.
Archives of environmental contamination and toxicology, 11(6), 699-702 (1982-11-01)
A Hirashima et al.
Bioorganic & medicinal chemistry, 8(3), 653-656 (2000-03-25)
Molecular interaction between enantiomeric fonofos oxon (O-ethyl S-phenyl ethylphosphonothiolate) and acetylcholinesterase (AChE) of Torpedo californica was evaluated by using the Cerius2 program. It was suggested that the difference in the inhibitory activity of the two enantiomers of fonofos oxon on
J F Fairchild et al.
Archives of environmental contamination and toxicology, 22(4), 375-379 (1992-05-01)
This study determined the acute and chronic toxicity of the organophosphate insecticide fonofos to standard freshwater aquatic organisms under laboratory conditions. Fonofos was acutely toxic to bluegill (Lepomis macrochirus), Daphnia (D. magna), and midge (Chironomous riparius) at 5.3, 2.7, and
Persistence and degradation of PP993 pyrethroid, fonofos, and chlorpyrifos in a Quebec cornfield's soil.
F A Elhag et al.
Bulletin of environmental contamination and toxicology, 42(2), 172-176 (1989-02-01)
W O Cook et al.
Veterinary and human toxicology, 27(4), 281-282 (1985-08-01)
An organophosphate toxicosis due to fonofos was diagnosed in an Iowa Dairy herd. Fonofos was detected in the feed source and in the rumen contents, liver, and kidney tissue of one cow that died acutely. Bulk tank milk contained detectable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service