Skip to Content
Merck
All Photos(1)

Key Documents

32349

Supelco

Heptachlor

PESTANAL®, analytical standard

Synonym(s):

1,4,5,6,7,8,8-Heptachloro-3a,4,7,7a-tetrahydro-4,7-methanoindene

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€161.00

€161.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€161.00

About This Item

Empirical Formula (Hill Notation):
C10H5Cl7
CAS Number:
Molecular Weight:
373.32
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

€161.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=C(Cl)[C@]2(Cl)C(Cl)(Cl)[C@@]1(Cl)[C@H]3[C@@H]2C(Cl)C=C3

InChI

1S/C10H5Cl7/c11-4-2-1-3-5(4)9(15)7(13)6(12)8(3,14)10(9,16)17/h1-5H/t3?,4?,5?,8-,9+/m1/s1

InChI key

FRCCEHPWNOQAEU-ZFBAKZBRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

E A Fendick et al.
Reviews of environmental contamination and toxicology, 111, 61-142 (1990-01-01)
The chlorinated cyclodiene heptachlor was registered in 1952 as an agricultural and domestic insecticide. By early 1984, registration for all purposes, except subterranean termite control and for limited use in the control of fire ants, had been cancelled. This restriction
Chlordane and heptachlor.
IARC monographs on the evaluation of carcinogenic risks to humans, 53, 115-175 (1991-01-01)
Hai Gen Zuo et al.
Environmental monitoring and assessment, 186(4), 2399-2412 (2013-12-18)
A new method has been developed to determine heptachlor and its metabolites heptachlor-exo-epoxide and heptachlor-endo-epoxide in pork. The pork samples were extracted with acetone-n-hexane (2:8, V:V) and cleaned up by gel permeation chromatography and florisil solid-phase extraction cartridge. The extract
Jianqiao Xu et al.
Environmental science & technology, 48(14), 8012-8020 (2014-06-17)
Bioconcentration factors (BCFs) measured in the laboratory are important for characterizing the bioaccumulative properties of chemicals entering the environment, especially the potential persistent organic pollutants (POPs), which can pose serious adverse effects on ecosystem and human health. Traditional lethal analysis
M D Reuber
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer, 7(3), 85-114 (1987-01-01)
Heptachlor and its metabolite heptachlor epoxide are unequivocally carcinogenic in rats and mice. The chemicals induced carcinomas of the liver, which were highly significant. There were neoplasms at other sites in rats. Neoplasms at all sites, as well as malignant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service