Skip to Content
Merck
All Photos(1)

Key Documents

32886

Supelco

4-tert-Octylphenol diethoxylate solution

10 μg/mL in acetone, analytical standard

Synonym(s):

2-{2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethoxy}ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€198.00

€198.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€198.00

About This Item

Empirical Formula (Hill Notation):
C18H30O3
CAS Number:
Molecular Weight:
294.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€198.00


Estimated to ship on18 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCO)cc1

InChI

1S/C18H30O3/c1-17(2,3)14-18(4,5)15-6-8-16(9-7-15)21-13-12-20-11-10-19/h6-9,19H,10-14H2,1-5H3

InChI key

LBCZOTMMGHGTPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sigma-Aldrich offers 4-tert-octylphenol diethoxylate solution (10 μg/mL in acetone) for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analyzes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service