Skip to Content
Merck
All Photos(1)

Key Documents

32894

Supelco

Nonylphenol monoethoxylate solution

50 μg/mL in acetone, technical, analytical standard

Synonym(s):

NP-1EO technical

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€213.00

€213.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€213.00

About This Item

Empirical Formula (Hill Notation):
C17H28O2
Molecular Weight:
264.40
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€213.00


Estimated to ship on23 May 2025


Request a Bulk Order

grade

analytical standard
technical

shelf life

limited shelf life, expiry date on the label

concentration

50 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nonylphenol monoethoxylates are a class of non-ionic surfactants, which are well-known precursors of estrogenic metabolic products found during wastewater treatment.[1]

Application

Nonylphenol monoethoxylate has been used as an analytical standard for the determination of the analyte in soils treated with sewage sludge by pressurized liquid extraction (PLE), followed by liquid chromatography-mass spectrometry (LC-MS).[1] It may be used as a reference standard for the analysis of the analyte in wastewater and sewage sludge by gas chromatography-mass spectrometry (GC-MS),[2] and in textile industry, waste water treatment plant effluents and surface waters by solid phase extraction (SPE) followed by LC-MS.[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of octylphenol, nonylphenol, alkylphenol ethoxylates and alcohol ethoxylates by pressurized liquid extraction and liquid chromatography-mass spectrometry in soils treated with sewage sludges.
Andreu V, et al.
The Science of the Total Environment, 378(1-2), 124-129 (2007)
LC-MS-MS analysis and occurrence of octyl- and nonylphenol, their ethoxylates and their carboxylates in Belgian and Italian textile industry, waste water treatment plant effluents and surface waters.
Loos R, et al.J
Chemosphere, 66(4), 690-699 (2007)
Simultaneous determination of the endocrine disrupting compounds nonylphenol, nonylphenol ethoxylates, triclosan and bisphenol A in wastewater and sewage sludge by gas chromatography-mass spectrometry.
Gatidou G, et al.
Journal of Chromatography A, 1138(1-2), 32-41 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service