Skip to Content
Merck
All Photos(1)

Key Documents

32800

Supelco

Nitarsone

VETRANAL®, analytical standard

Synonym(s):

4-Nitrobenzenearsonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6AsNO5
CAS Number:
Molecular Weight:
247.04
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

O[As](O)(=O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C6H6AsNO5/c9-7(10,11)5-1-3-6(4-2-5)8(12)13/h1-4H,(H2,9,10,11)

InChI key

FUUFQLXAIUOWML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fa Zhao et al.
Journal of chromatography. A, 1573, 48-58 (2018-09-10)
A sensitive, simple and rapid method for the simultaneous determination of eleven arsenic species has been developed. A high performance liquid chromatography-inductively coupled plasma mass spectrometry (HPLC-ICP-MS) technique was used for the analysis of eleven arsenic species in less than
Jian Chen et al.
Environmental science & technology, 53(11), 6182-6191 (2019-05-07)
Organoarsenical biotransformations are important components of the global cycling of arsenic. Roxarsone (3-nitro-4-hydroxybenzenearsenate or Rox(V)) and nitarsone (4-nitrobenzene arsenate or Nit(V)) are synthetic aromatic organoarsenicals used in the poultry industry as additives to prevent coccidiosis and improve feed efficiency. Here
C F Duffy et al.
Avian diseases, 49(3), 423-425 (2005-10-29)
Histomoniasis (histomonosis, infectious enterohepatitis, or blackhead) is a disease of turkeys on litter or range caused by the protozoan Histomonas meleagridis, a parasite of worms, primarily spread in feces, in Heterakis gallinarum (cecal worm) eggs, or in Eisenia foetida (earthworms).
Daniel Sánchez-Rodas et al.
Analytical and bioanalytical chemistry, 385(7), 1172-1177 (2006-03-22)
A rapid extraction procedure has been developed for speciation of arsenic in chicken tissue. Water, methanol-water (1:1), and methanol-chloroform (1:1) were tested as extraction media. Individual use of an ultrasonic bath, a microwave oven, or an ultrasonic probe was not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service