Skip to Content
Merck
All Photos(2)

Key Documents

A9258

Sigma-Aldrich

p-Arsanilic acid

Synonym(s):

4-Aminobenzenearsonic acid, 4-Aminophenylarsonic acid, 4-Arsanilic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€46.00

€46.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€46.00

About This Item

Empirical Formula (Hill Notation):
C6H8AsNO3
CAS Number:
Molecular Weight:
217.05
Beilstein:
1102334
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€46.00


Check Cart for Availability

Request a Bulk Order

form

powder

mp

≥300 °C (lit.)

SMILES string

Nc1ccc(cc1)[As](O)(O)=O

InChI

1S/C6H8AsNO3/c8-6-3-1-5(2-4-6)7(9,10)11/h1-4H,8H2,(H2,9,10,11)

InChI key

XKNKHVGWJDPIRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Marianna Czaplicka et al.
Environmental science and pollution research international, 22(21), 16927-16935 (2015-06-26)
The paper presents the kinetics and proposed pathways photodegradation and photooxidation of p-arsanilic acid, in a neutral environment by ozone and hydrogen peroxide. The results showed that in a neutral environment, photoozonation process was characterized by the highest decomposition rate
Han Young Eom et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1006, 151-157 (2015-11-10)
A simple and sensitive derivatization method using toluene-3,4-dithiol as a derivatization reagent for the simultaneous analysis of seven arsenic compounds (roxarsone, nitarsone, p-arsanilic acid, o-arsanilic acid, phenylarsonic acid, phenylarsine oxide, and mono-methylarsonic acid) in chicken muscle was developed and validated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service