Skip to Content
Merck
All Photos(1)

Key Documents

32641

Supelco

S-Metolachlor Metabolite CGA 368208

PESTANAL®, analytical standard

Synonym(s):

[(2-Ethyl-6-methyl-phenyl)-carbamoyl]-methanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€504.00

€504.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€504.00

About This Item

Empirical Formula (Hill Notation):
C11H14NNaO4S
CAS Number:
Molecular Weight:
279.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€504.00


Estimated to ship on21 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[Na+].CCc1cccc(C)c1NC(=O)CS([O-])(=O)=O

InChI

1S/C11H15NO4S.Na/c1-3-9-6-4-5-8(2)11(9)12-10(13)7-17(14,15)16;/h4-6H,3,7H2,1-2H3,(H,12,13)(H,14,15,16);/q;+1/p-1

InChI key

WWFKJLARXKHBKN-UHFFFAOYSA-M

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
S-Metolachlor Metabolite CGA 368208 may be used as a pestanal standard for the estimation of the analyte in surface water and groundwater samples using direct injection liquid chromatography-electrospray ionization-mass spectrometry (LC-ESI-MS) operating in multiple reaction monitoring (MRM).[1]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A multimethod for the determination of 150 pesticide metabolites in surface water and groundwater using direct injection liquid chromatography-mass spectrometry.
Reemtsma, et al.
Journal of Chromatography A, 1271(1), 95-104 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service