Skip to Content
Merck
All Photos(1)

Key Documents

32559

Supelco

Ethyl 4-hydroxybenzoate-ring-13C6 solution

50 μg/mL in acetone, analytical standard

Synonym(s):

4-Hydroxybenzoic acid ethyl ester-ring-13C6 solution, Ethyl paraben-ring-13C6 solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€412.00

€412.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€412.00

About This Item

Empirical Formula (Hill Notation):
13C6C3H10O3
Molecular Weight:
172.13
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€412.00


Estimated to ship on21 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

50 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

mass shift

M+5

storage temp.

−20°C

SMILES string

CCOC(=O)[13c]1[13cH][13cH][13c](O)c([13cH]1)C#CC#CC#CC#CC#CC#C

InChI key

UFWYPRJFXXFGOS-HUXPIBMOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service