Skip to Content
Merck
All Photos(2)

Key Documents

32521

Supelco

Azimsulfuron

PESTANAL®, analytical standard

Synonym(s):

N-{[(4,6-Dimethoxy-2-pyrimidinyl)amino]carbonyl}-1-methyl-4-(2-methyl-H-tetrazol-5-yl)-1H-pyrazole-5-sulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€604.00

€604.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€604.00

About This Item

Empirical Formula (Hill Notation):
C13H16N10O5S
CAS Number:
Molecular Weight:
424.40
Beilstein:
8284859
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€604.00


Estimated to ship on16 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1cc(OC)nc(NC(=O)NS(=O)(=O)c2c(cnn2C)-c3nnn(C)n3)n1

InChI

1S/C13H16N10O5S/c1-22-11(7(6-14-22)10-18-21-23(2)19-10)29(25,26)20-13(24)17-12-15-8(27-3)5-9(16-12)28-4/h5-6H,1-4H3,(H2,15,16,17,20,24)

InChI key

MAHPNPYYQAIOJN-UHFFFAOYSA-N

General description

Azimsulfuron is a sulfonylurea based pesticide.[1]

Application

Azimsulfuron may be used as an analytical reference standard for the determination of the pesticide in:
  • Paddy field water by direct injection using liquid chromatography-hybrid triple quadrupole-linear ion trap-mass spectrometry (LC-QqLIT-MS).[1]
  • Tap and leaching water samples by solid-phase extraction (SPE), followed by LC coupled to tandem mass spectrometry (LC-MS/MS).[2]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trace analysis of sulfonylurea herbicides in water samples by solid-phase extraction and liquid chromatography-tandem mass spectrometry.
Fenoll J, et al.
Talanta, 101(30), 273-282 (2012)
Molecular basis of Cyperus difformis cross-resistance to ALS-inhibiting herbicides.
S Ntoanidou et al.
Pesticide biochemistry and physiology, 127, 38-45 (2016-01-30)
J D Obourn et al.
Toxicology and applied pharmacology, 146(1), 116-126 (1997-09-23)
A8947 is a member of the sulfonyl urea class of compounds and is the active ingredient in a commercial broad leaf herbicide. This compound has been shown to produce pancreatic hypertrophy in rats, mice, and dogs. The objectives of this
HPLC-UV and HPLC-MSn multiresidue determination of amidosulfuron, azimsulfuron, nicosulfuron, rimsulfuron, thifensulfuron methyl, tribenuron methyl and azoxystrobin in surface waters.
Polati S, et al.
Analytica Chimica Acta, 579(2), 146-151 (2006)
Trace analysis of pesticides in paddy field water by direct injection using liquid chromatography-quadrupole-linear ion trap-mass spectrometry
Pareja L, et al.
Journal of Chromatography A, 1218(30), 4790-4798 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service