Skip to Content
Merck
All Photos(1)

Key Documents

32240

Sigma-Aldrich

Diethylene glycol monoethyl ether acetate

≥99.0% (GC)

Synonym(s):

2-(2-Ethoxyethoxy)ethyl acetate, Ethyldiglycol acetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€78.60

€78.60


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€78.60

About This Item

Linear Formula:
C2H5OCH2CH2OCH2CH2CO2CH3
CAS Number:
Molecular Weight:
176.21
Beilstein:
1764643
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€78.60


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥99.0% (GC)

refractive index

n20/D 1.421 (lit.)
n20/D 1.421

bp

218-219 °C (lit.)

density

1.011 g/mL at 25 °C (lit.)

functional group

ester
ether

SMILES string

CCOCCOCCOC(C)=O

InChI

1S/C8H16O4/c1-3-10-4-5-11-6-7-12-8(2)9/h3-7H2,1-2H3

InChI key

FPZWZCWUIYYYBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service