Skip to Content
Merck
All Photos(3)

Key Documents

32208

Sigma-Aldrich

Diethylene glycol dimethyl ether

puriss., absolute, over molecular sieve (H2O ≤0.005%), ≥99.5% (GC)

Synonym(s):

2-Methoxyethyl ether, Bis(2-methoxyethyl) ether, Dimethyldiglycol, ‘Diglyme’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OCH2CH2)2O
CAS Number:
Molecular Weight:
134.17
Beilstein:
1736101
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

vapor density

4.6 (vs air)

vapor pressure

3 mmHg ( 20 °C)

grade

absolute
puriss.

Assay

≥99.5% (GC)

autoignition temp.

370 °F

quality

over molecular sieve (H2O ≤0.005%)

expl. lim.

17.4 %

refractive index

n20/D 1.408 (lit.)
n20/D 1.408

bp

162 °C (lit.)

mp

−64 °C (lit.)

density

0.944 g/mL at 20 °C (lit.)
0.939 g/mL at 25 °C (lit.)

SMILES string

COCCOCCOC

InChI

1S/C6H14O3/c1-7-3-5-9-6-4-8-2/h3-6H2,1-2H3

InChI key

SBZXBUIDTXKZTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethylene glycol dimethyl ether (diglyme), a less volatile liquid, is widely employed as an anhydrous solvent. Various physical properties (density, refractive index, speed of sound and shear viscosity) of its mixtures with ethyl acetate, methyl benzoate, ethyl benzoate or diethyl succinate have been evaluated at various temperature ranges.

Application

Diethylene glycol dimethyl ether (diglyme) was used for the immobilization of biological compounds onto a gold surface via dextran chemistry. It was employed as solvent for the functionalization of carbon nano fibers, via the Diels Alder reaction. It may be used in the preparation of linear poly(glycidol) (PG), via anionic polymerization. It may be used as solvent for the extraction of tattoo colorants and their degradation products from various media by recovery experiments and HPLC-diode array detector technology.

recommended

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Repr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F M Fernandes et al.
Journal of nanoscience and nanotechnology, 7(10), 3514-3518 (2008-03-12)
This paper reports functionalization of CNF via a Diels-Alder addition reaction and the characterization of the obtained materials. The functionalization was assessed by a calorimetric technique (DSC) and the morphology of CNF modified materials was characterized by scanning electron microscopy
Biocompatible and degradable nanogels via oxidation reactions of synthetic thiomers in inverse miniemulsion.
Groll J, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 47(20), 5543-5549 (2009)
Ilaria Mannelli et al.
Biosensors & bioelectronics, 22(6), 803-809 (2006-04-20)
Two different surface chemistries have been studied for the development of surface plasmon resonance imaging (SPRI) based DNA microarray affinity sensors: (1) 11-mercaptoundecanoic acid-poly(ethylenimine) (MUA-PEI) and (2) dextran procedures. The MUA-PEI method consists of assembling a multilayer on the basis
Densities, refractive indices, speeds of sound, and shear viscosities of diethylene glycol dimethyl ether with ethyl acetate, methyl benzoate, ethyl benzoate, and diethyl succinate in the temperature range from 298.15 to 318.15 K.
Aminabhavi TM, et al.
Journal of Chemical and Engineering Data, 39(2), 251-260 (1994)
Eva Engel et al.
Analytical chemistry, 78(18), 6440-6447 (2006-09-15)
Tattooing is a widespread process of puncturing pigments into skin, whereas the resulting concentration inside the skin remains unknown. Many tattoo colorants are organic pigments, such as azo pigments, manufactured for other uses. To remove tattoos from skin, laser pulses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service