Skip to Content
Merck
All Photos(1)

Documents

31418

Supelco

Dextran from Leuconostoc mesenteroides

analytical standard, for GPC, Mw 12,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

mol wt

Mn ~8,000
Mp ~10,000
Mw ~12,000

analyte chemical class(es)

oligosaccharides

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.50

application(s)

food and beverages

format

neat

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, with a linear backbone of α-linked D-glucopyranosyl repeating units. Dextrans are found as bacterial extracellular polysaccharides. They are synthesized from sucrose by Leuconostoc mesenteroides and Lactobacillus brevis. Bacteria employ dextran in biofilm formation or as a protective coating to evade host phagocytes in the case of pathogenic bacteria.
Dextran from Leuconostoc mesenteroides (Mw: 12,000) may be used as an analytical standard to calibrate the column for gel permeation chromatography (GPC).

Application

Use of dextrans as long and hydrophilic spacer arms improves the performance of immobilized proteins acting on macromolecules.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Luisa Miranda-Vilela et al.
Journal of biomedical nanotechnology, 9(7), 1261-1271 (2013-08-06)
This work aimed to test a dextran-functionalized magnetic fluid (DexMF) sample in mediating magnetohyperthermia to treat an advanced clinical Ehrlich-solid-tumor, to verify the effects of oral antioxidant administration of pequi-oil on this treatment and to investigate the potential of these
Elizabeth M Bradshaw et al.
Nature neuroscience, 16(7), 848-850 (2013-05-28)
In our functional dissection of the CD33 Alzheimer's disease susceptibility locus, we found that the rs3865444(C) risk allele was associated with greater cell surface expression of CD33 in the monocytes (t50 = 10.06, P(joint) = 1.3 × 10(-13)) of young
Ngalle Eric Mbua et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(25), 10207-10212 (2013-06-05)
Niemann-Pick type C (NPC) disease is characterized by impaired cholesterol efflux from late endosomes and lysosomes and secondary accumulation of lipids. Although impaired trafficking of individual glycoproteins and glycolipids has been noted in NPC cells and other storage disorders, there
David Needham et al.
Faraday discussions, 161, 515-534 (2013-06-29)
This paper describes how we have used material science, physical chemistry, and some luck, to design a new thermal-sensitive liposome (the low temperature sensitive liposome (LTSL)) that responds at clinically attainable hyperthermic temperatures releasing its drug in a matter of
Hideto Katsuta et al.
Investigative ophthalmology & visual science, 54(6), 4102-4108 (2013-05-23)
To elucidate the role of signals mediated by EphB4 receptor tyrosine kinase and its transmembrane ephrinB2 ligand in corneal lymphatic capillaries. To detect expression of ephrinB2 and EphB4 in mouse corneas, immunohistochemistry of flat-mount corneas from 6- to 10-week-old wild-type

Related Content

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service