Skip to Content
Merck

Skip To

90475

Blue Dextran

average mol wt 10,000

Synonym(s):

Blue Dextran 10, Dextran blue from Leuconostoc mesenteroides

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
€ 208,00

About This Item

CAS Number:
UNSPSC Code:
12352116
NACRES:
NA.32
MDL number:

€ 208,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

form

powder

mol wt

average mol wt 10,000

loss

≤10% loss on drying

capacity

0.01-0.08 mmol/g loading

storage temp.

−20°C


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
90008D477272762
biological source

synthetic

biological source

synthetic

biological source

-

biological source

-

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

loss

≤10% loss on drying

loss

≤10% loss on drying

loss

-

loss

≤10% loss on drying

mol wt

average mol wt 10,000

mol wt

average mol wt 5,000

mol wt

~2,000,000

mol wt

average mol wt 500,000


Still not finding the right product?

Explore all of our products under Blue Dextran


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters