Skip to Content
Merck
All Photos(1)

Key Documents

27698

Sigma-Aldrich

Colominic acid sodium salt from Escherichia coli

≥95% (NANA Sodium salt)

Synonym(s):

Poly[2,8-(N-acetylneuraminic acid sodium salt)]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C11H16NNaO8)n
CAS Number:
MDL number:
UNSPSC Code:
12352201

Assay

≥95% (NANA Sodium salt)

optical activity

[α]20/D +15.5±1.0°, c = 2.5% in H2O

solubility

H2O: 25 mg/mL at 20 °C, clear, colorless

antibiotic activity spectrum

viruses

storage temp.

2-8°C

InChI

1S/C11H20NO7.3Na/c1-6(15)12-10-7(16)4-9(18-3-2-13)19-11(10)8(17)5-14;;;/h3,7-11,13-14,16-17H,2,4-5H2,1H3,(H,12,15);;;/q;3*+1

InChI key

IPYNPWIUCZLARC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Substrate for the assay of polysialic acid depolymerase; Assay of α-2#8594;8 sialidase; Structure and chemistry of colominic acid

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polysialic acid depolymerase.
B Kwiatkowski et al.
Methods in enzymology, 138, 786-792 (1987-01-01)
S. Okada et al.
Perspect. Inherited Metab. Dis. 1980, 4, 145-145 (1981)
THE STRUCTURE AND CHEMISTRY OF COLOMINIC ACID.
E J MCGUIRE et al.
Biochemistry, 3, 247-251 (1964-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service