Skip to Content
Merck
All Photos(1)

Key Documents

27095

Supelco

Chromoionophore VI

Selectophore

Synonym(s):

4′,5′-Dibromofluorescein octadecyl ester, ETH 7075

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H46Br2O5
CAS Number:
Molecular Weight:
742.58
Beilstein:
8247765
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

mp

214-216 °C

SMILES string

CCCCCCCCCCCCCCCCCCOC(=O)c1ccccc1C2=C3C=CC(=O)C(Br)=C3Oc4c(Br)c(O)ccc24

InChI

1S/C38H46Br2O5/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-19-26-44-38(43)28-21-18-17-20-27(28)33-29-22-24-31(41)34(39)36(29)45-37-30(33)23-25-32(42)35(37)40/h17-18,20-25,41H,2-16,19,26H2,1H3

InChI key

CIRQTSPKAXLQLF-UHFFFAOYSA-N

General description

Chromoionophore VI (ETH 7075) is a lipophilic pH indicator and is generally used during the fabrication of optical sensors.[1]
Visit our Sensor Applications portal to learn more.

Application

ETH 7075 was used for the preparation of optical membrane, used for the determination of glucose.[2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Lipophilized pH-indicator used as chromoionophore in chloride sensors[3]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of glucose using a coupled-enzymatic reaction with new fluoride selective optical sensing polymeric film coated in microtiter plate wells.
Abd-Rabboh
Talanta, 72 (3), 1129-1133 (2007)
Highly selective optical fluoride ion sensor with submicromolar detection limit based on aluminum (III) octaethylporphyrin in thin polymeric film.
Badr IHA and Meyerhoff ME.
Journal of the American Chemical Society, 127 (15), 5318-5319 (2005)
S.S. Tan et al.
Analytica Chimica Acta, 255, 35-35 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service