Skip to Content
Merck
All Photos(2)

Key Documents

216720

Sigma-Aldrich

4′,5′-Dibromofluorescein

Dye content 95 %

Synonym(s):

Solvent Red 72

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€55.50

€55.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€55.50

About This Item

Empirical Formula (Hill Notation):
C20H10Br2O5
CAS Number:
Molecular Weight:
490.10
Colour Index Number:
45370:1
Beilstein:
57189
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€55.50


Check Cart for Availability

Request a Bulk Order

Assay

>95% (TLC)

form

powder

composition

Dye content, 95%

mp

270-273 °C (lit.)

λmax

450 nm

ε (extinction coefficient)

≥5000 at 445-451 nm
≥50000 at 227-233 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc2c(Oc3c(Br)c(O)ccc3C24OC(=O)c5ccccc45)c1Br

InChI

1S/C20H10Br2O5/c21-15-13(23)7-5-11-17(15)26-18-12(6-8-14(24)16(18)22)20(11)10-4-2-1-3-9(10)19(25)27-20/h1-8,23-24H

InChI key

ZDTNHRWWURISAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4′,5′-Dibromofluorescein (DBF) is a fluorone dye. Fluorone dyes are hydroxyl-xanthene dyes (homologues of fluorescein) and have been the most widely used class of organic fluorophores for labeling and sensing biomolecules. DBF is routinely used as a ligand for studying the properties of protein using spectral analysis.

Application

4′,5′-Dibromofluorescein has been used to stain proteins in 1-D and 2-D sodium dodecyl sulfatepolyacrylamide gel electrophoresis SDS-PAGE. This method is rapid, economical, and highly sensitive in comparison with other proteomic research staining methods.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gzona Bajraktari-Sylejmani et al.
European journal of drug metabolism and pharmacokinetics, 45(6), 809-815 (2020-07-15)
Citrus flavonoids are not only components of daily nutrition, they are also promoted as dietary supplements and are important ingredients in traditional medicines. Interactions of flavonoids with synthetic drugs represent an often neglected issue. We therefore investigated in vitro whether
Marc Le Vée et al.
Journal of biochemical and molecular toxicology, 33(10), e22379-e22379 (2019-08-01)
The interactions of six neonicotinoid pesticides and one neonicotinoid metabolite with drug transporters have been characterized in vitro. Acetamiprid, clothianidin, imidacloprid, nitenpyram, thiacloprid and its metabolite thiacloprid amide, and thiamethoxam, each used at 100 µM, did not impair activity of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service