Skip to Content
Merck
All Photos(1)

Key Documents

227358

Sigma-Aldrich

3,4,5,6-Tetrabromophenolsulfonephthalein

Dye content 95 %

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€614.00

€614.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€614.00

About This Item

Empirical Formula (Hill Notation):
C19H10Br4O5S
CAS Number:
Molecular Weight:
669.96
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€614.00


Estimated to ship on21 May 2025


Request a Bulk Order

form

powder

Quality Level

composition

Dye content, 95%

visual transition interval

6.6-8.2, yellow to purple

λmax

426 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc(cc1)C2(OS(=O)(=O)c3c(Br)c(Br)c(Br)c(Br)c23)c4ccc(O)cc4

InChI

1S/C19H10Br4O5S/c20-14-13-18(17(23)16(22)15(14)21)29(26,27)28-19(13,9-1-5-11(24)6-2-9)10-3-7-12(25)8-4-10/h1-8,24-25H

InChI key

ARVZQGSXIKSAAY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service