Skip to Content
Merck
All Photos(3)

Key Documents

236047

Sigma-Aldrich

Tetrabromophenol Blue sodium salt

Dye content 85 %, ≥84.5% (HPLC)

Synonym(s):

3,3′,3′′,4,5,5′,5′′,6-Octabromophenolsulfonphthalein sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€78.70
5 G
€258.00

€78.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€78.70
5 G
€258.00

About This Item

Empirical Formula (Hill Notation):
C19H5Br8NaO5S
CAS Number:
Molecular Weight:
1007.53
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€78.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥84.5% (HPLC)

form

powder

composition

Dye content, 85%

technique(s)

titration: suitable

color

brown to dark brown

λmax

610 nm

ε (extinction coefficient)

≥12000 at 298-304 nm in H2O at 0.005 g/L
≥55000 at 607-613 nm in H2O at 0.005 g/L
≥59000 at 222-228 nm in H2O at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Oc1c(Br)cc(cc1Br)C(=C2\C=C(Br)C(=O)C(Br)=C2)\c3c(Br)c(Br)c(Br)c(Br)c3S([O-])(=O)=O

InChI

1S/C19H6Br8O5S.Na/c20-7-1-5(2-8(21)17(7)28)11(6-3-9(22)18(29)10(23)4-6)12-13(24)14(25)15(26)16(27)19(12)33(30,31)32;/h1-4,28H,(H,30,31,32);/q;+1/p-1

InChI key

GFMOPDHPAXHAQX-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service