20980-U
Hamilton® syringe, 7000 series, knurled hub
7105, volume 5 μL, needle size 24 ga (bevel tip), needle L 70 mm (2.75 in.)
Synonym(s):
Precision Syringe
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
bevel tip stainless steel needle (point style 2)
needle size
24 ga (bevel tip)
feature
Sample contained in needle (no dead volume)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 88011
needle L
70 mm (2.75 in.)
volume
5 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Rapid injection of minute volumes
- Eliminates dead volume by placing the sample in the needle
- Leak-proof at injection pressures up to 6000 psi
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service