Skip to Content
Merck
All Photos(1)

Key Documents

18602

Supelco

Mineral oil standard mixture Type A and B for EN ISO 9377-2

analytical standard, ~10 mg/mL in heptane (exact content on the reference material certificate)

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€56.80

€56.80


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
2 ML
€56.80

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€56.80


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Agency

EN 9377-2
ISO

shelf life

limited shelf life, expiry date on the label

concentration

~10 mg/mL in heptane (exact content on the reference material certificate)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.686 g/mL at 20 °C

application(s)

environmental
petroleum

format

multi-component solution

General description

Mineral oil standard mixture is mostly used to determine mineral oil content in water, soil and sludge matrices.

Application

It may be used as reference material in determination of total hydrocarbon index from water samples, which involves direct coupling between stir membrane extraction and infrared spectroscopy.[1]
Mineral oil standard mixture type A and B for EN ISO 9377-2 has been used as a reference matrix spiking solution in the determination of total petroleum hydrocarbons in samples of sea sediments using gas chromatography coupled with flame ionization detection (GC-FID).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of Total Hydrocarbons and Alkyl PAHs in Sediments from Apsheron Peninsula in Caspian Sea.
Ibadov AN and Suleymanov AB
Biological and Chemical Research, 111-111 (2015)
Sensitive in-surface infrared monitoring coupled to stir membrane extraction for the selective determination of total hydrocarbon index in waters.
Alcudia-Leon MC
Analytical and Bioanalytical Chemistry, 398(3), 1427-1433 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service