Skip to Content
Merck

Skip To

49574

Extraction Medium Stock Solution

acc. to DIN EN ISO 9377-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Check Cart for Availability
€ 93,20

About This Item

UNSPSC Code:
41113319
EC Number:
205-563-8
NACRES:
NA.21

€ 93,20


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

application(s)

environmental

Physical form

contains n-Decane und n-Tetracontane as internal standard, in n-Heptane

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
5006585170651730
grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

Quality Level

100

Quality Level

300

Quality Level

100

Quality Level

100

application(s)

environmental

application(s)

petroleum

application(s)

environmental
petroleum

application(s)

petroleum


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

24.8 °F - closed cup

flash_point_c

-4 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Quali sono le concentrazioni in mg/L di n decano e n tetracontano?

    1 answer
    1. The concentrations of n-decane and n-tetracontane may vary slightly from batch to batch. According to DIN EN ISO 9377-2, the prescribed concentration is approximately 20 mg/L in heptane for n-tetracontane and approximately 15 mg/L in heptane for n-decane. This standard is primarily used for identifying the gas chromatography retention time window for hydrocarbons between n-decane and n-tetracontane, and the exact concentrations are not quantified for each batch.

      Helpful?

Reviews

No rating value

Active Filters