Skip to Content
Merck
All Photos(1)

Key Documents

17124

Millipore

Algae Culture Broth

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€159.00

€159.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€159.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

€159.00


Check Cart for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

ammonium chloride, 0.05 g/L
calcium chloride, 0.058 g/L
dipotassium phosphate, 0.25 g/L
ferric chloride, 0.003 g/L
magnesium sulfate, 0.513 g/L
sodium nitrate, 1 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

agriculture
environmental
food and beverages
life science and biopharma
water monitoring

microbiology

suitability

algae

Application

For the isolation and cultivation of algae from soil, water and sewage.

Preparation Note

Suspend 1.87 g in 1 litre distilled water. Boil to dissolve the medium completely. Dispense as desired. Sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Dears, is this medium used also for cyanobacteria?

    1 answer
    1. As mentioned under suitability in the PROPERTIES section of the product page, this product is listed as being suitable for use in the culture of cyanobacteria. In addition, there are two other products that are specifically formulated for the culture of these organisms. To review each option please see the links below.
      C3061, Cyano BG-11 Medium
      https://www.sigmaaldrich.com/product/sigma/c3061

      73818, BG11 Broth
      https://www.sigmaaldrich.com/product/sial/73816

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service