Skip to Content
Merck
All Photos(2)

Key Documents

1.07298

Supelco

Hydrogen peroxide solution

30%, Suprapur®

Synonym(s):

Hydrogen peroxide solution, Perhydrol

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€155.00
500 ML
€233.00
1 L
€388.00

€155.00


Estimated to ship on30 March 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€155.00
500 ML
€233.00
1 L
€388.00

About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304
NACRES:
NB.21
concentration:
30%
grade:
for inorganic trace analysis
Agency:
suitable for SM 2310
suitable for SM 2320
pH:
2-4 (20 °C in H2O)

€155.00


Estimated to ship on30 March 2025


Request a Bulk Order

Product Name

Hydrogen peroxide 30%, Suprapur®

grade

for inorganic trace analysis

Quality Level

Agency

suitable for SM 2310
suitable for SM 2320

vapor pressure

18 hPa ( 20 °C)
23.3 mmHg ( 30 °C)

form

liquid

concentration

30%

pH

2-4 (20 °C in H2O)

bp

107 °C/1013 hPa

mp

-26 °C

density

1.11 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Versatile Platinum Nanoparticles-Decorated Phage Nanozyme Integrating Recognition, Bacteriolysis, and Catalysis Capabilities for On-Site Detection of Foodborne Pathogenic Strains Vitality Based on Bioluminescence/Pressure Dual-Mode Bioassay: This innovative study utilizes 30% hydrogen peroxide as a critical component in the development of a phage nanozyme. This application is instrumental for rapid on-site detection of pathogenic bacteria in food safety testing, showcasing hydrogen peroxide′s role in enhancing public health monitoring (You et al., 2024).

  • Integrated physiological, transcriptomic and metabolomic analyses provide insights into phosphorus-mediated cadmium detoxification in Salix caprea roots: In this environmental science study, hydrogen peroxide is used to study stress responses in plants, offering a comprehensive understanding of its utility in plant physiology research and potential implications for improving phytoremediation technologies (Li et al., 2024).

Linkage

Replaces: 7298-2; 7298-4; 7298

Analysis Note

Assay (manganometric): ≥ 30.0 %
Free acid (as H₂SO₄): ≤ 30 ppm
Total nitrogen (N): ≤ 3 ppm
Chloride (Cl): ≤ 300 ppb
Nitrate (NO₃): ≤ 500 ppb
Phosphate (PO₄): ≤ 100 ppb
Sulphate (SO₄): ≤ 100 ppb
Ag (Silver): ≤ 0.5 ppb
Al (Aluminium): ≤ 1.0 ppb
As (Arsenic): ≤ 1.0 ppb
Au (Gold): ≤ 0.5 ppb
B (Boron): ≤ 0.5 ppb
Ba (Barium): ≤ 0.5 ppb
Be (Beryllium): ≤ 0.5 ppb
Bi (Bismuth): ≤ 0.5 ppb
Ca (Calcium): ≤ 5 ppb
Cd (Cadmium): ≤ 0.5 ppb
Co (Cobalt): ≤ 0.5 ppb
Cr (Chromium): ≤ 0.5 ppb
Cu (Copper): ≤ 0.5 ppb
Fe (Iron): ≤ 1.0 ppb
Ga (Gallium): ≤ 0.5 ppb
Ge (Germanium): ≤ 0.5 ppb
Hg (Mercury): ≤ 1 ppb
In (Indium): ≤ 0.5 ppb
K (Potassium): ≤ 1.0 ppb
Li (Lithium): ≤ 0.5 ppb
Mg (Magnesium): ≤ 1.0 ppb
Mn (Manganese): ≤ 0.5 ppb
Mo (Molybdenum): ≤ 0.5 ppb
Na (Sodium): ≤ 1.0 ppb
Ni (Nickel): ≤ 0.5 ppb
Pb (Lead): ≤ 0.5 ppb
Pd (Palladium): ≤ 0.5 ppb
Pt (Platinum): ≤ 0.5 ppb
Sb (Antimony): ≤ 0.5 ppb
Sn (Tin): ≤ 0.5 ppb
Sr (Strontium): ≤ 0.5 ppb
Ti (Titanium): ≤ 1 ppb
Tl (Thallium): ≤ 0.5 ppb
V (Vanadium): ≤ 0.5 ppb
Zn (Zinc): ≤ 1.0 ppb
Zr (Zirconium): ≤ 0.5 ppb
The actual values are subject to unavoidable systematic
variations in this concentration range.

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuğba Ekiz Yılmaz et al.
Acta histochemica, 122(2), 151470-151470 (2019-12-10)
Cyclophosphamide (CYP) is one of the alkylating chemotherapeutic agents and its adverse effects on folliculogenesis in the ovary are well-known due to the previous scientific research on this topic. Magnesium has various effects in organisms, including catalytic functions on the

Articles

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

A 4-step comprehensive ICP-MS workflow including sample homogenization, with standard addition calibration method, to determine heavy metals in Cannabis sativa.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service