Skip to Content
Merck
All Photos(1)

Key Documents

161993

Sigma-Aldrich

p-Toluenesulfonic acid monohydrate

reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3H · H2O
CAS Number:
Molecular Weight:
190.22
Beilstein:
3568023
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor density

5.9 (vs air)

Assay

98%

mp

103-106 °C (lit.)

SMILES string

[H]O[H].Cc1ccc(cc1)S(O)(=O)=O

InChI

1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2

InChI key

KJIFKLIQANRMOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

comparable product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detailed characterization of p-toluenesulfonic acid monohydrate as a convenient, recoverable, safe, and selective catalyst for alkylation of the aromatic nucleus.
Mahindaratne MPD and Wimalasena K.
The Journal of Organic Chemistry, 63(9), 2858-2866 (1998)
Makoto Ueno et al.
The Journal of organic chemistry, 68(16), 6424-6426 (2003-08-05)
alpha-Tosyloxyketones and alpha-tosyloxyaldehydes were directly prepared from alcohols by treatment with iodosylbenzene and p-toluenesulfonic acid monohydrate in good yields. This method can be used for the direct preparation of thiazoles, imidazoles, and imidazo[1,2-a]pyridines from alcohols in good to moderate yields
Solvent-Free Reductive Amination: An Organic Chemistry Experiment.
Goldstein SW and Cross AV.
Journal of Chemical Education (2015)
The crystal and molecular structure of 4-methyl sulfonic acid (p-toluenesulfonic acid) monohydrate, C7H8SO3-. H3O+, an oxonium salt.
Arora SK and Sundaralingam M.
Acta Crystallographica Section B, Structural Science, 27(7), 1293-1298 (1971)
Hans H Adomat et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 902, 84-95 (2012-07-24)
Androgens are key mediators of prostate development and function, a role that extends to the development of prostate diseases such as benign prostatic hyperplasia (BPH) and prostate cancer. In prostate, DHT is the major androgen and reduction and glucuronidation are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service