Skip to Content
Merck
All Photos(1)

Key Documents

16174

Sigma-Aldrich

SODOSIL RA 21

acid pre-rinse and rinsing solution with phosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

UBA Number:
08050017
UNSPSC Code:
12352000
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

pH

1-3 (20 °C, 1%)

density

1.40-1.46 g/mL at 20 °C

Application

SODOSIL RA 21 is a sterilization and decontamination kit, generally employed in disinfecting lab containers or surfaces used for downstream scientific studies utilizing cell culture, and associated techniques.[1]

Legal Information

SODOSIL is a trademark of Sigma-Aldrich Chemie GmbH

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple cleaning and fluorescent staining protocol for recent and fossil diatom frustules.
Friedrichs L.
Diatom research, 28(3), 317-327 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service