Skip to Content
Merck
All Photos(1)

Key Documents

16173

Sigma-Aldrich

SODOSIL RA 15

liquid, alkaline cleaner free of phosphates, low foaming for rinsing apparatus

Synonym(s):

Silica-based Surfactant

Sign Into View Organizational & Contract Pricing

Select a Size

5 L
€131.00
35 KG
Inquire

€131.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
5 L
€131.00
35 KG
Inquire

About This Item

UBA Number:
08050015
UNSPSC Code:
12352000
NACRES:
NA.22

€131.00


Estimated to ship on27 May 2025


Request a Bulk Order

Quality Level

pH

12-14 (20 °C, direct)

density

1.28-1.34 g/mL at 20 °C

General description

SODOSIL RA 15 is a nonionic, phosphate-free detergent kit for cleaning lab containers and surfaces used in various scientific studies.[1]

Application

The additon of 30-50ml cleaner per washing cycle (about 10lt); pH-value of the washing liquor (0,3%): 11.4, contains sodium hydroxide

Legal Information

SODOSIL is a trademark of Sigma-Aldrich Chemie GmbH

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A candidate reference method for the determination of magnesium in serum
Kulpmann WR, et al.
Journal of Clinical Chemistry and Clinical Biochemistry (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service