Skip to Content
Merck
All Photos(2)

Key Documents

14593

Supelco

4-Methyl-1H-benzotriazole

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€63.20

€63.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€63.20

About This Item

Empirical Formula (Hill Notation):
C7H7N3
CAS Number:
Molecular Weight:
133.15
Beilstein:
119183
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€63.20


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

140-146 °C

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cc1cccc2[nH]nnc12

InChI

1S/C7H7N3/c1-5-3-2-4-6-7(5)9-10-8-6/h2-4H,1H3,(H,8,9,10)

InChI key

CMGDVUCDZOBDNL-UHFFFAOYSA-N

General description

4-Methyl-1H-benzotriazole is an additive in formulated aircraft deicing and anti-icing fluids (ADAFs) mixture.[1]

Application

It was used as analytical standard for a study demonstrated to determine benzotriazoles and benzothiazoles in human urine samples using LC- ESI(+)MS/MS.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sospeter N Njeru et al.
Oncogene, 39(7), 1445-1465 (2019-10-30)
Aneuploidy can instigate tumorigenesis. However, mutations in genes that control chromosome segregation are rare in human tumors as these mutations reduce cell fitness. Screening experiments indicate that the knockdown of multiple classes of genes that are not directly involved in
Alexandros G Asimakopoulos et al.
Analytical chemistry, 85(1), 441-448 (2012-12-06)
Benzotriazole (BTR) and benzothiazole (BTH) derivatives are used in a wide variety of industrial and consumer products and have been reported to occur in the environment. Owing to a lack of analytical methods, human exposure to BTR and BTH is
Devon A Cancilla et al.
Environmental toxicology and chemistry, 22(1), 134-140 (2002-12-31)
This paper presents the results of a number of field and laboratory studies to evaluate the environmental impact of aircraft deicing and anti-icing fluids (ADAFs) on aquatic systems. Both 5-methyl-1H-benzotriazole (5-MeBt) and 4-methyl-1H-benzotriazole (4-MeBt), known additives to ADAFs, were found
Jiufeng Li et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1070, 70-75 (2017-11-02)
Benzotriazole (BTR) and benzothiazole (BTH) derivatives are extensively applied in industrial processes and consumer products, and are thus frequently detected in the environmental matrices. Due to their potential estrogenic effects reported in animal studies, the assessment of human exposure to
Attilio Naccarato et al.
Journal of chromatography. A, 1338, 164-173 (2014-03-19)
This work proposes a new approach for the simultaneous determination of benzothiazoles, benzotriazoles and benzosulfonamides in different environmental matrices and human urine, using solid-phase microextraction coupled with gas-chromatography-triple quadrupole mass spectrometry (SPME-GC-QqQMS). The analytes object of this investigation have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service