Skip to Content
Merck
All Photos(2)

Documents

196304

Sigma-Aldrich

5-Methyl-1H-benzotriazole

98%

Synonym(s):

5-Methyl-1,2,3-benzotriazole, 5-Methylbenzotriazole, 5-Tolyltriazole, 6-Methyl-1,2,3-benzotriazole, 6-Methyl-1H-benzotriazole, 6-Tolyltriazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N3
CAS Number:
Molecular Weight:
133.15
Beilstein:
116658
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

bp

210-212 °C/12 mmHg (lit.)

mp

80-82 °C (lit.)

SMILES string

Cc1ccc2[nH]nnc2c1

InChI

1S/C7H7N3/c1-5-2-3-6-7(4-5)9-10-8-6/h2-4H,1H3,(H,8,9,10)

InChI key

LRUDIIUSNGCQKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Methyl-1H-benzotriazole is an active component of aircraft deicing and anti-icing fluid. It prevents the corrosion of copper and brass in a variety of corrosive environments.

Application

5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corrosion inhibition of copper by tolyltriazole.
Walker R.
Corrosion (Houston, TX, United States), 32(8), 339-341 (1976)
Carmen Domínguez et al.
Journal of chromatography. A, 1230, 117-122 (2012-02-18)
Benzothiazoles (BTHs) and benzotriazoles (BTRs) belong to a high production volume chemicals widely used in both industrial and household applications. Since they are recalcitrant to biodegradation, they are widespread in the environment. However, the BTHs and BTRs determination in environmental
Devon A Cancilla et al.
Environmental toxicology and chemistry, 22(1), 134-140 (2002-12-31)
This paper presents the results of a number of field and laboratory studies to evaluate the environmental impact of aircraft deicing and anti-icing fluids (ADAFs) on aquatic systems. Both 5-methyl-1H-benzotriazole (5-MeBt) and 4-methyl-1H-benzotriazole (4-MeBt), known additives to ADAFs, were found
Rozanna Avagyan et al.
Environmental science and pollution research international, 22(8), 5842-5849 (2014-10-25)
Textiles play an important role in our daily life, and textile production is one of the oldest industries. In the manufacturing chain from natural and/or synthetic fibers to the final clothing products, the use of many different chemicals is ubiquitous.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service