Skip to Content
Merck
All Photos(1)

Key Documents

13464-M

Sigma-Aldrich

Sodium sulfate

puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 KG
€74.50

€74.50


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
2.5 KG
€74.50

About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.0-100.5% (calc. to the dried substance)
form:
crystalline

€74.50


Estimated to ship on16 April 2025


Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

grade

puriss.

Assay

99.0-100.5% (calc. to the dried substance)

form

crystalline

quality

meets analytical specification of Ph. Eur., BP, USP

impurities

acidity or alkalinity, complies
residual solvents, complies
≤0.001% heavy metals (as Pb)
≤0.04% free alkali (as NaOH)
≤0.05% free acid (as H2SO4)

loss

≤0.5% loss on drying, 130 °C

pH

5.2-8 (20 °C, 50 g/L)

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.02%

cation traces

As: ≤0.0002%
Ca: ≤0.005%
Fe: ≤0.005%
Mg: ≤0.005%
Zn: ≤0.03%

suitability

complies for appearance of solution

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium sulfate (Na2SO4) can be obtained directly via precipitation from a saturated solution of Na2SO4 at room temperature (20°C). Its crystallization and growth rates at room temperature (20°C) under various conditions have been investigated.[1] Sodium sulfate can be produced during Leblanc process, from sodium chloride and sulfuric acid.[2]

Application

Sodium sulfate (Na2SO4) may be employed as a reagent for the precipitation of globulins during the estimation of proteins in blood.[3]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service