Skip to Content
Merck
All Photos(2)

Key Documents

1.06637

Supelco

Sodium sulfate

anhydrous, coarse granules for analysis EMSURE® ACS

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€48.70
1 KG
€74.90
25 KG
€824.00

€48.70


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€48.70
1 KG
€74.90
25 KG
€824.00

About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-820-9
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
200 g/L

€48.70


Estimated to ship on30 May 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

≥99.0% (acidimetric)

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

loss

≤0.5% loss on ignition, 800 °C

pH

5.2-8.0 (20 °C, 50 g/L in H2O)

mp

884 °C (lit.)

solubility

200 g/L

density

2.68 g/mL at 25 °C (lit.)

bulk density

1400‑1600 kg/m3

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%

cation traces

As: ≤0.0001%
Ca: ≤0.01%
Fe: ≤0.0005%
K: ≤0.002%
Mg: ≤0.001%
N: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Removal of Sodium Sulfate from Iron Phosphate: Sodium sulfate is also involved in methods for removing impurities from chemical compounds. A recent study focused on developing a method to remove sodium sulfate from iron phosphate, an essential process in the production of high-purity materials for various industrial applications. This research highlights the importance of sodium sulfate in refining chemical processes and improving the quality of industrial products (Li et al., Advances in Analytical Chemistry, 2022).

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Identity: passes test
Insoluble matter: ≤ 0.01 %
pH-value (5 %; water): 5.2 - 9.2
Chloride (Cl): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.001 %
Total nitrogen (N): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
As (Arsenic): ≤ 0.0001 %
Ca (Calcium): ≤ 0.01 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.002 %
Mg (Magnesium): ≤ 0.001 %
Loss on ignition (800 °C): ≤ 0.5 %
Particle size (250-2000 µm): ≥ 85 %
Corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

High resolution separations of low molecular weight heparin-daletparin and enoxaparin can be achived using Zenix SEC-100 columns.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service