Skip to Content
Merck
All Photos(1)

Key Documents

11523

Supelco

Lanthanum Standard for ICP

TraceCERT®, 1 g/L La in nitric acid (nominal concentration)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€54.20

€54.20


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€54.20

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€54.20


Estimated to ship on18 May 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1 g/L La in nitric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Lanthanum ICP standard can be used in the determination of lanthanum in Raphidocelis subcapitata microalgae by inductively coupled plasma mass spectrometry (ICP-MS).[1]

Analysis Note

1 g/L La in 2% nitric acid, prepared with high purity La2O3, HNO3, and water

Other Notes

For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long-term multi-endpoint exposure of the microalga Raphidocelis subcapitata to lanthanum and cerium
Siciliano A, et al.
The Science of the Total Environment, 790, 148229-148229 (2021)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service