Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
product line
TraceCERT®
grade
certified reference material, TraceCERT®
concentration
1 g/L Ga in nitric acid (nominal concentration)
technique(s)
ICP: suitable
application(s)
industrial qc
pharmaceutical
format
single component solution
General description
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Search & download your certificate here.
Application
Analysis Note
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) ICP: suitable | technique(s) ICP: suitable | technique(s) ICP: suitable | technique(s) ICP: suitable |
| grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® |
| format single component solution | format single component solution | format single component solution | format single component solution |
| product line TraceCERT® | product line TraceCERT® | product line TraceCERT® | product line TraceCERT® |
| application(s) industrial qc | application(s) industrial qc | application(s) industrial qc | application(s) industrial qc |
| concentration 1 g/L Ga in nitric acid (nominal concentration) | concentration 1 g/L Pd in hydrochloric acid (nominal concentration) | concentration 1 g/L Ce in nitric acid (nominal concentration) | concentration 1 g/L Pt in hydrochloric acid (nominal concentration) |
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
supp_hazards
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




