Skip to Content
Merck
All Photos(3)

Key Documents

318736

Sigma-Aldrich

Bromocresol Purple solution

0.04 wt. % in H2O

Synonym(s):

5′,5′′-Dibromo-o-cresolsulfonephthalein sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€31.30
500 ML
€105.00

€31.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€31.30
500 ML
€105.00

About This Item

Empirical Formula (Hill Notation):
C21H15Br2NaO5S
CAS Number:
Molecular Weight:
562.20
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€31.30


Check Cart for Availability

Request a Bulk Order

form

liquid

concentration

0.04 wt. % in H2O

pH

5.2-6.8, yellow-green to purple

density

0.991 g/mL at 25 °C

λmax

590 nm

ε (extinction coefficient)

≥60000 at 584-590 nm in 10 mM NaOH

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Cc1cc(cc(Br)c1O)\C(=C2/C=C(C)C(=O)C(Br)=C2)c3ccccc3S([O-])(=O)=O

InChI

1S/C21H16Br2O5S.Na/c1-11-7-13(9-16(22)19(11)24)21(14-8-12(2)20(25)17(23)10-14)15-5-3-4-6-18(15)29(26,27)28-21;/h3-10,24-25H,1-2H3;/q;+1/p-1

InChI key

WNOIORUCCYNQPE-UHFFFAOYSA-M

General description

Bromocresol purple (BCP) is an organic dye[1] and a pH indicator.[2] It changes from yellow (pH 5.2) to purple (6.8).[2] BCP reacts with ammonia to yield ammonium salt. Hence, it can be used as an ammonia sensing probe.[1]

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Acid-Base Indicators (2007)
Optical fiber ammonia sensing probes using reagent immobilized porous silica coating as transducers
Tao S, et al.
Sensors and Actuators B, Chemical, 115(1), 158-163 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service