Skip to Content
Merck
All Photos(2)

Documents

11223

Supelco

3,3′,5,5′-Tetrabromobisphenol A

analytical standard

Synonym(s):

4,4′-Isopropylidenebis(2,6-dibromophenol), TBBPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C[C6H2(Br)2OH]2
CAS Number:
Molecular Weight:
543.87
Beilstein:
1889048
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97% (HPLC)
96.5-103.5% (wt.)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

177-183 °C
178-181 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC(C)(c1cc(Br)c(O)c(Br)c1)c2cc(Br)c(O)c(Br)c2

InChI

1S/C15H12Br4O2/c1-15(2,7-3-9(16)13(20)10(17)4-7)8-5-11(18)14(21)12(19)6-8/h3-6,20-21H,1-2H3

InChI key

VEORPZCZECFIRK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,3′,5,5′-Tetrabromobisphenol A is a brominated derivative of bisphenol A commonly used as a flame retardant. It finds applications in a variety of consumer products ranging from fabrics to plastics and electronics. Its removal from the aquatic environment using the environmental-friendly chitosan (CS)/poly(vinyl alcohol) (PVA) nanofibrous membranes is reported.

Application

3,3′,5,5′-Tetrabromobisphenol A may be used as an analytical reference standard for the quantification of the analyte in air samples and sediment/sludge samples using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An environmentally-friendly enzyme-based nanofibrous membrane for 3, 3?, 5, 5?-tetrabromobisphenol removal
Xu R, et al.
Royal Society of Chemistry Advances, 5(79), 64091-64097 (2015)
Terminal elimination half-lives of the brominated flame retardants TBBPA, HBCD, and lower brominated PBDEs in humans.
Geyer, Harald J., et al.
Organohalogen compounds, 66, 3867-3872 (2004)
Zhiyong Xie et al.
Analytica chimica acta, 584(2), 333-342 (2007-03-28)
A simple and effective method has been developed for analysis of the flame retardant tetrabromobisphenol A (TBBPA) in environmental samples by using modified soxhlet extraction in combination with silica gel clean-up, derivatization with silylation reagent and gas chromatography-mass spectrometry (GC-MS)
Courtney C Carignan et al.
Environmental science & technology, 46(21), 12146-12153 (2012-09-25)
Tetrabromobisphenol-A (TBBP-A) and hexabromocyclododecanes (HBCDs) are brominated flame retardants that have been found in human milk and serum throughout the world, but have received comparatively little attention in the United States. The objective of this study is to determine concentrations
Guillaume ten Dam et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 898, 101-110 (2012-05-18)
Since the EFSA enquired a call for data for TBBPA and HBCD in 2009, the analytical determination of these compounds in food became of regulatory interest. Therefore, a method for the simultaneous determination of TBBPA and the three major HBCD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service