Skip to Content
Merck

Skip To

50877

Bisphenol B

analytical standard

Synonym(s):

2,2-Bis(4-hydroxyphenyl)butane, 4,4′-sec-Butylidenediphenol, NSC 1775

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 83,20

About This Item

Empirical Formula (Hill Notation):
C16H18O2
CAS Number:
Molecular Weight:
242.31
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
201-025-1
MDL number:

€ 83,20


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

SMILES string

OC(C=C1)=CC=C1C(C)(CC)C2=CC=C(O)C=C2

InChI

1S/C16H18O2/c1-3-16(2,12-4-8-14(17)9-5-12)13-6-10-15(18)11-7-13/h4-11,17-18H,3H2,1-2H3

InChI key

HTVITOHKHWFJKO-UHFFFAOYSA-N

General description

Bisphenol B is a congener of bisphenol A, used primarily for the manufacturing of phenolic resins and is also a potent food contaminant.
This compound is listed in the SVHC (Substances of very high concern) candidate list of ECHA (European Chemicals Agency)

Application

Bisphenol B may be used as a reference standard for the determination of bisphenol B in liquid food matrices and powdered infant formulae and human urine samples by using dispersive liquid-liquid microextraction (DLLME) with in situ derivatization coupled with heart-cutting gas chromatography-mass spectrometry (GC-MS).

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
924275145390477
grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

format

neat

format

neat

format

neat

format

neat

assay

≥98.0% (HPLC)

assay

≥95.0% (HPLC)

assay

≥98.0% (HPLC)

assay

≥99.0% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label


pictograms

Exclamation mark

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - ED ENV 1 - ED HH 1 - Eye Irrit. 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

77-40-7

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters