Skip to Content
Merck
All Photos(1)

Key Documents

1.01193

Sigma-Aldrich

MagPrep® Silica Particles

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€51.50
50 ML
€2,670.00
500 ML
€24,630.00
1000 ML
€49,250.00

€51.50


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€51.50
50 ML
€2,670.00
500 ML
€24,630.00
1000 ML
€49,250.00

About This Item

UNSPSC Code:
12352401
NACRES:
NA.21

€51.50


Estimated to ship on20 May 2025


Request a Bulk Order

density

1.04 g/cm3 at 20 °C

Quality Level

storage temp.

2-8°C

Analysis Note

Appearance: black suspension
Magnetic Response: 1 - 20 s
Settling time: ≥ 5 min
RNases: not detectable
Fe (Iron): ≤ 5 ppm
Particlesurface (BET): 14 - 25 m²/g
For research use only

Legal Information

MAGPREP is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover how MagPrep® Silica particles meet essential needs for DNA and RNA nucleic acid purification protocols.

Questions

1–2 of 2 Questions  
  1. What are the dimensions of the particles in product 1.01193, including the size of the magnetic core and the thickness of the silica coating?

    1 answer
    1. MagPrep Silica particles have a diameter of 50-150 nm and are suspended in 50mM MES buffer at pH 5.5.

      Helpful?

  2. Please provide clarification on the pH during the lysis and binding steps for the MagPrep products

    1 answer
    1. Different lysis methods may have specific pH requirements. The pH is a secondary factor in nucleic acid binding to the silica, with the salts in the buffer being the primary driver of the silica's negative charge. While optimizing the pH can enhance yields, the recommended protocols should be ideal.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service