Skip to Content
Merck
All Photos(1)

Key Documents

81108

Supelco

Micro particles based on silicon dioxide

size: 2 μm

Synonym(s):

Silica dioxide microsphere (2 μm), Micro silicon (2 μm), Beads based on silicon dioxide, microsize, Silicon dioxide beads

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€159.00
10 ML
€297.00

€159.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€159.00
10 ML
€297.00

About This Item

MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.25

€159.00


Estimated to ship on14 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

form

aqueous suspension

concentration

5% (solids)

particle size

2 μm std dev ≤0.2 μm

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silicon dioxide based microparticles are suitable for profiling particle size distribution (PSD) of test samples.
The particle diameter of 2 μm is evaluated using Coulter Multisizer.

Application

Used to qualify and monitor the performance of particle size equipment.
Also used as a calibration standard to:
  • assess 3D particle tracking ability of optical diffraction tomography instrument for optically trapped silica beads[1]
  • synthesize inverse patchy colloids (IPCs) for phase-behavior studies of heterogeneously charged particles[2]

Features and Benefits

  • suitable for routine instrument calibration monitoring, testing and corrections
  • particle size traceable to Community Bureau of Reference (BCR) standards
  • available in 5 mL and 10 mL pack size as neat samples

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous 3D visualization and position tracking of optically trapped particles using optical diffraction tomography
Kim K, et al.
Optica, 2(4), 343-346 (2015)
Simple method for the synthesis of inverse patchy colloids
van Oostrum PL, et al.
Journal of Physics. Condensed Matter : An Institute of Physics Journal, 27(23), 234105-234105 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service