Skip to Content
Merck
All Photos(2)

Documents

1.05423

Supelco

Ammonia solution

28-30%, for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Ammonium hydroxide solution, Ammonia water, Ammonium hydroxide solution, Ammonia aqueous, Ammonia water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein:
3587154
MDL number:
UNSPSC Code:
12352402
NACRES:
NA.21

product name

Ammonia solution 28-30%, for analysis EMSURE® ACS,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

635 hPa ( 20 °C)

product line

EMSURE®

form

liquid

expl. lim.

27 %

concentration

28.0-30.0% NH4OH

impurities

≤0.020 ppm Ag (Silver)
≤0.020 ppm Ga (Gallium)
≤0.020 ppm In (Indium)
≤0.020 ppm Li (Lithium)
≤0.050 ppm Ba (Barium)
≤0.050 ppm Cd (Cadmium)
≤0.050 ppm Co (Cobalt)
≤0.050 ppm Cr (Chromium)
≤0.050 ppm Mn (Manganese)
≤0.050 ppm Mo (Molybdenum)
≤0.050 ppm Ni (Nickel)
≤0.050 ppm Pb (Lead)
≤0.050 ppm Tl (Thallium)
≤0.100 ppm Au (Gold)
≤0.100 ppm Bi (Bismuth)
≤0.100 ppm Cu (Copper)
≤0.100 ppm Fe (Iron)
≤0.100 ppm Mg (Magnesium)
≤0.100 ppm Pt (Platinum)
≤0.100 ppm Sn (Tin)
≤0.100 ppm Sr (Strontium)
≤0.100 ppm Ti (Titanium)
≤0.100 ppm Zn (Zinc)
≤0.2 ppm Sulphide (S)
≤0.3 ppm Chloride (Cl)
≤0.5 ppm Heavy metals (as Pb)
≤0.5 ppm Phosphate (PO4)
≤0.500 ppm Al (Aluminium)
≤0.500 ppm As (Arsenic)
≤0.500 ppm Ca (Calcium)
≤0.500 ppm K (Potassium)
≤0.500 ppm Na (Sodium)
≤10 ppm Carbonate (as CO2)
≤10 ppm Non volatile matter
≤2 ppm Nitrate (NO3)
≤2 ppm Pyridine and related substances
≤2 ppm Sulphate (SO4)
≤5 ppm Silicate (as SiO2)
≤5 ppm Substances reducing potassium permanganate

SMILES string

[NH4+].[OH-]

ign. residue

≤10 ppm

pH

>12 (20 °C in H2O, strongly alkaline)

bp

32 °C

mp

-72 °C

density

0.9 g/mL at 25 °C (lit.)
0.892-0.910 (relative density (d 20/20))

storage temp.

2-30°C

InChI

1S/H3N.H2O/h1H3;1H2

InChI key

VHUUQVKOLVNVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

Due to quality and safety aspects Ammonia Solution will no longer be offered in glass bottles but solely in HDPE bottles.

Please note the following replacements:
1054231000 (1 L glass bottle) replaced by 1054231011 (1 L HDPE bottle)
1054232500 (2.5 L glass bottle) replaced by 1054232511 (2.5 L HDPE bottle)

More information about our ergonomic and unbreakable HDPE bottles can be found here: www.SigmaAldrich.com/hdpe-bottle

Analysis Note

Assay (acidimetric, NH₃): 28.0 - 30.0 %
Identity: passes test
Appearance: passes test
Appearance of solution: passes test
Relative Density (d 20/20): 0.892 - 0.910
Carbonate (as CO₂): ≤ 10 ppm
Chloride (Cl): ≤ 0.3 ppm
Nitrate (NO₃): ≤ 2 ppm
Phosphate (PO₄): ≤ 0.5 ppm
Heavy metals (as Pb): ≤ 0.5 ppm
Silicate (as SiO₂): ≤ 5 ppm
Sulphate (SO₄): ≤ 2 ppm
Sulphide (S): ≤ 0.2 ppm
Ag (Silver): ≤ 0.020 ppm
Al (Aluminium): ≤ 0.500 ppm
As (Arsenic): ≤ 0.500 ppm
Au (Gold): ≤ 0.100 ppm
Ba (Barium): ≤ 0.050 ppm
Bi (Bismuth): ≤ 0.100 ppm
Ca (Calcium): ≤ 0.500 ppm
Cd (Cadmium): ≤ 0.050 ppm
Co (Cobalt): ≤ 0.050 ppm
Cr (Chromium): ≤ 0.050 ppm
Cu (Copper): ≤ 0.100 ppm
Fe (Iron): ≤ 0.100 ppm
Ga (Gallium): ≤ 0.020 ppm
In (Indium): ≤ 0.020 ppm
K (Potassium): ≤ 0.500 ppm
Li (Lithium): ≤ 0.020 ppm
Mg (Magnesium): ≤ 0.100 ppm
Mn (Manganese): ≤ 0.050 ppm
Mo (Molybdenum): ≤ 0.050 ppm
Na (Sodium): ≤ 0.500 ppm
Ni (Nickel): ≤ 0.050 ppm
Pb (Lead): ≤ 0.050 ppm
Pt (Platinum): ≤ 0.100 ppm
Sn (Tin): ≤ 0.100 ppm
Sr (Strontium): ≤ 0.100 ppm
Ti (Titanium): ≤ 0.100 ppm
Tl (Thallium): ≤ 0.050 ppm
Zn (Zinc): ≤ 0.100 ppm
Pyridine and related substances: ≤ 2 ppm
Substances Reducing Permanganate (ACS): passes test
Substances reducing potassium permanganate: ≤ 5 ppm
Residue after ignition: ≤ 10 ppm
Non volatile matter: ≤ 10 ppm

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This study is aimed at developing and validating an LC-ESI-ion trap-MS/MS method for identification and quantification of ethyl glucuronide (EtG) and ethyl sulfate (EtS) as ethanol biomarkers for urine analysis.

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service