Skip to Content
Merck
All Photos(1)

Documents

09164

Supelco

(+)-Dihydrocarvone, mixture of isomers

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.2 (vs air)

vapor pressure

0.06 mmHg ( 20 °C)

Assay

≥98% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.471 (lit.)

bp

87-88 °C/6 mmHg (lit.)

density

0.929 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC1CCC(CC1=O)C(C)=C

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)10(11)6-9/h8-9H,1,4-6H2,2-3H3

InChI key

AZOCECCLWFDTAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dihydrocarvone, an oxygenated monoterpene, is used in the synthesis of several products, including chiral bicyclic phenols, hydroxy ketone 7, lucinone, etc. Dihydrocarvone is also a potential insect repellent known to be effective against the rice weevil Sitophilus oryzae (L), and an inhibitor of bacterial and fungal growth.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(+)-Occidentalol: Absolute Stereostructure and Total Synthesis
Amano Y and Heathcock CH
Canadian Journal of Chemistry, 50(3), 340-345 (1972)
Application of the tandem Stryker reduction--aldol cyclization strategy to the asymmetric synthesis of lucinone
Chiu P, et al.
Tetrahedron Letters, 42(24), 4091-4093 (2001)
(R)-(-)-carvone and (1R, 4R)-trans-(+)-dihydrocarvone from poiretia latifolia vogel
Porto C, et al.
Journal of the Brazilian Chemical Society, 21(5), 782-786 (2010)
Aromatic annulation: Two new methods for the synthesis of chiral bicyclic phenols
Corey EJ and Palani A
Tetrahedron Letters, 38(14), 2397-2400 (1997)
A Convenient Procedure for the Preparation of Dihydrocarvone.
Raucher S and Hwang K-J.
Synthetic Communications, 10(2), 133-137 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service