Skip to Content
Merck
All Photos(1)

Key Documents

W356506

Sigma-Aldrich

D-Dihydrocarvone, mixture of isomers

≥97%, FCC, FG

Synonym(s):

p-Menth-8-en-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

W356506-SAMPLE-K
€47.40
100 G
€73.00
1 KG
€305.00
5 KG
€1,020.00

€47.40


Estimated to ship on28 March 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W356506-SAMPLE-K
€47.40
100 G
€73.00
1 KG
€305.00
5 KG
€1,020.00

About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
FEMA Number:
3565
EC Number:
Council of Europe no.:
11703
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.128
NACRES:
NA.21
Organoleptic:
caraway; herbaceous; minty; woody; ethereal; spicy; camphoraceous
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens

€47.40


Estimated to ship on28 March 2025


Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥97%

optical activity

[α]20/D +16°, neat

refractive index

n20/D 1.472 (lit.)

bp

87-88 °C/6 mmHg (lit.)

density

0.926 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

caraway; herbaceous; minty; woody; ethereal; spicy; camphoraceous

SMILES string

CC1CCC(CC1=O)C(C)=C

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)10(11)6-9/h8-9H,1,4-6H2,2-3H3

InChI key

AZOCECCLWFDTAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service