Skip to Content
Merck
All Photos(2)

Key Documents

06561

Sigma-Aldrich

Formic acid triethylamine complex 5:2

≥99.0% (NT)

Synonym(s):

Triethylammonium formate 2:5

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€105.00
250 ML
€414.00

€105.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
€105.00
250 ML
€414.00

About This Item

Linear Formula:
5HCOOH · 2N(CH2CH3)3
CAS Number:
Molecular Weight:
86.50
Beilstein:
4150472
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€105.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.0% (NT)

refractive index

n20/D 1.428

functional group

amine

SMILES string

OC=O.OC=O.OC=O.OC=O.OC=O.CCN(CC)CC.CCN(CC)CC

InChI

1S/2C6H15N.5CH2O2/c2*1-4-7(5-2)6-3;5*2-1-3/h2*4-6H2,1-3H3;5*1H,(H,2,3)

InChI key

NLOIHFJXBNFDSF-UHFFFAOYSA-N

Related Categories

Application

Reductant for iridium-catalyzed greener amine synthesis by transfer hydrogenation of imines.

A Versatile Catalyst for Reductive Amination by Transfer Hydrogenation

Other Notes

Activated form of formic acid used for reduction reactionscitation
Activated form of formic acid used for reduction reactions[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Wagner
Angewandte Chemie (International Edition in English), 82, 73-73 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service