Skip to Content
Merck
All Photos(1)

Key Documents

04249

Sigma-Aldrich

Potassium phosphate tribasic monohydrate

90%

Synonym(s):

tert-Potassium phosphate monohydrate, Tripotassium phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€68.00
1 KG
€102.00

€68.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
500 G
€68.00
1 KG
€102.00

About This Item

Empirical Formula (Hill Notation):
K3O4P · H2O
CAS Number:
Molecular Weight:
230.28
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
grade:
for analytical purposes

€68.00


Estimated to ship on01 June 2025


Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

90%

impurities

≤0.002% heavy metals (as Pb)

loss

≤13% loss on ignition, 800 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Fe: ≤20 mg/kg

SMILES string

O.[K+].[K+].[K+].[O-]P([O-])([O-])=O

InChI

1S/3K.H3O4P.H2O/c;;;1-5(2,3)4;/h;;;(H3,1,2,3,4);1H2/q3*+1;;/p-3

InChI key

RMNIZOOYFMNEJJ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Potassium phosphate tribasic can be used in Suzuki-Miyaura cross-coupling reactions of γ-boronated esters with aromatic halides to synthesize regioselectively substituted amino esters with the retention of configurations.[1]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regio-and Stereocontrolled Access to ?-Boronated Unsaturated Amino Esters and Derivatives from (Z)-Alkenyl 1, 2-Bis (boronates)
Sridhar T, et al.
The Journal of Organic Chemistry, 79(2), 783-789 (2013)
Damião Pergentino de Sousa et al.
Natural product communications, 10(11), 1893-1896 (2016-01-12)
Aromatic plants produce volatile substances with high therapeutic potential. In view of the need for new respiratory and cardiovascular system pharmacological agents, the present study reports on the spasmolytic activity of two enantiomers of carvone and limonene, constituents of essential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service